Skip to Content
MilliporeSigma
Tetrabutylammonium hydroxide 40% in water, EMPLURA®

Tetrabutylammonium hydroxide

Tetrabutylammonium hydroxide 40% in water, EMPLURA®
Synonym(s):
Tetrabutylammonium hydroxide solution
Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47

Products

Brand
Feature
Formula Weight
Product Line
Showing 1-1 of 1
Compare
Product Number
Description
Pricing