Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.94955

Sigma-Aldrich

Tetrabutylammonium hydroxide

new

40% in water, EMPLURA®

Synonym(s):

Tetrabutylammonium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47
Beilstein/REAXYS Number:
3571228
MDL number:

product line

EMPLURA®

concentration

40% in water

storage temp.

room temp

SMILES string

[OH-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1

InChI key

VDZOOKBUILJEDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service