MilliporeSigma
  • Enzymatic synthesis of methyl derivatives of L-tryptophan selectively labeled with hydrogen isotopes.
Enzymatic synthesis of methyl derivatives of L-tryptophan selectively labeled with hydrogen isotopes.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2018-04-02)
Elżbieta Winnicka, Marianna Kańska
ABSTRACT
We report the enzymatic synthesis of the derivatives of L-tryptophan methylated in indole moiety and labeled with deuterium and tritium in the 2-position of side chain. For kinetic studies twelve isotopomers, i.e., 1'-methyl-[2-2H]-, 1'-methyl-[2-3H]-, 1'-methyl-[2-2H/3H]-, 2'-methyl-[2-2H]-, 2'-methyl-[2-3H]-, 2'-methyl-[2-2H/3H]-, 5'-methyl-[2-2H]-, 5'-methyl-[2-3H]-, 5'-methyl-[2-2H/3H]-, 7'-methyl-[2-2H]-, 7'-methyl-[2-3H]-, and 7'-methyl-[2-2H/3H]-L-tryptophan are obtained by the enzymatic coupling of the appropriate methylated indole moiety with S-methyl-L-cysteine catalyzed by the enzyme tryptophanase.
Sigma-Aldrich
S-Methyl-L-cysteine, substrate for methionine sulfoxide reductase A