MilliporeSigma
  • Hypomethylation-Linked Activation of PLCE1 Impedes Autophagy and Promotes Tumorigenesis through MDM2-Mediated Ubiquitination and Destabilization of p53.
Hypomethylation-Linked Activation of PLCE1 Impedes Autophagy and Promotes Tumorigenesis through MDM2-Mediated Ubiquitination and Destabilization of p53.
Cancer research (2020-02-19)
Yunzhao Chen, Huahua Xin, Hao Peng, Qi Shi, Menglu Li, Jie Yu, Yanxia Tian, Xueping Han, Xi Chen, Yi Zheng, Jun Li, Zhihao Yang, Lan Yang, Jianming Hu, Xuan Huang, Zheng Liu, Xiaoxi Huang, Hong Zhou, Xiaobin Cui, Feng Li
ABSTRACT
Esophageal squamous cell carcinoma (ESCC) is one of the deadliest malignant diseases. Multiple studies with large clinic-based cohorts have revealed that variations of phospholipase C epsilon 1 (PLCE1) correlate with esophageal cancer susceptibility. However, the causative role of PLCE1 in ESCC has remained elusive. Here, we observed that hypomethylation-mediated upregulation of PLCE1 expression was implicated in esophageal carcinogenesis and poor prognosis in ESCC cohorts. PLCE1 inhibited cell autophagy and suppressed the protein expression of p53 and various p53-targeted genes in ESCC. Moreover, PLCE1 decreased the half-life of p53 and promoted p53 ubiquitination, whereas it increased the half-life of mouse double minute 2 homolog (MDM2) and inhibited its ubiquitination, leading to MDM2 stabilization. Mechanistically, the function of PLCE1 correlated with its direct binding to both p53 and MDM2, which promoted MDM2-dependent ubiquitination of p53 and subsequent degradation in vitro. Consequently, knockdown of PLCE1 combined with transfection of a recombinant adenoviral vector encoding wild-type p53 resulted in significantly increased levels of autophagy and apoptosis of esophageal cancer in vivo. Clinically, the upregulation of PLCE1 and mutant p53 protein predicted poor overall survival of patients with ESCC, and PLCE1 was positively correlated with p53 in ESCC cohorts. Collectively, this work identified an essential role for PLCE1- and MDM2-mediated ubiquitination and degradation of p53 in inhibiting ESCC autophagy and indicates that targeting the PLCE1-MDM2-p53 axis may provide a novel therapeutic approach for ESCC. SIGNIFICANCE: These findings identify hypomethylation-mediated activation of PLCE1 as a potential oncogene that blocks cellular autophagy of esophageal carcinoma by facilitating the MDM2-dependent ubiquitination of p53 and subsequent degradation. GRAPHICAL ABSTRACT: http://cancerres.aacrjournals.org/content/canres/80/11/2175/F1.large.jpg.
Sigma-Aldrich
MISSION® esiRNA, targeting human PLCE1
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder