MilliporeSigma
  • Frizzled 1 and Wnt1 as new potential therapeutic targets in the traumatically injured spinal cord.
Frizzled 1 and Wnt1 as new potential therapeutic targets in the traumatically injured spinal cord.
Cellular and molecular life sciences : CMLS (2020-01-05)
Pau González, Carlos González-Fernández, Yolanda Campos-Martín, Manuela Mollejo, Melissa Carballosa-Gautam, Alexander Marcillo, Michael Norenberg, Francisco Javier Rodríguez
ABSTRACT
Despite the experimental evidence pointing to a significant role of the Wnt family of proteins in physiological and pathological rodent spinal cord functioning, its potential relevance in the healthy and traumatically injured human spinal cord as well as its therapeutic potential in spinal cord injury (SCI) are still poorly understood. To get further insight into these interesting issues, we first demonstrated by quantitative Real-Time PCR and simple immunohistochemistry that detectable mRNA expression of most Wnt components, as well as protein expression of all known Wnt receptors, can be found in the healthy human spinal cord, supporting its potential involvement in human spinal cord physiology. Moreover, evaluation of Frizzled (Fz) 1 expression by double immunohistochemistry showed that its spatio-temporal and cellular expression pattern in the traumatically injured human spinal cord is equivalent to that observed in a clinically relevant model of rat SCI and suggests its potential involvement in SCI progression/outcome. Accordingly, we found that long-term lentiviral-mediated overexpression of the Fz1 ligand Wnt1 after rat SCI improves motor functional recovery, increases myelin preservation and neuronal survival, and reduces early astroglial reactivity and NG2+ cell accumulation, highlighting the therapeutic potential of Wnt1 in this neuropathological situation.
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sudan Black B, certified by the BSC
Sigma-Aldrich
Anti-Serotonin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 (Phos. and Non-Phos.) antibody produced in mouse, clone N52, ascites fluid
Sigma-Aldrich
Milli-Mark® Pan Neuronal Marker, Milli-Mark Pan Neuronal Marker is an antibody targeting the Pan Neuronal Marker protein, validated for use in ICC, IHC, IF & IHC.
Sigma-Aldrich
Anti-NeuN (rabbit monoclonal) Antibody, clone 27-4, clone 27-4, from rabbit
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%