MilliporeSigma
  • Rapid loss of blood-brain barrier P-glycoprotein activity through transporter internalization demonstrated using a novel in situ proteolysis protection assay.
Rapid loss of blood-brain barrier P-glycoprotein activity through transporter internalization demonstrated using a novel in situ proteolysis protection assay.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2010-07-16)
Brian T Hawkins, Robert R Rigor, David S Miller
ABSTRACT
Blood-brain barrier (BBB) P-glycoprotein activity is rapidly reduced by vascular endothelial growth factor (VEGF) acting via Src and by tumor necrosis factor-alpha acting via protein kinase C (PKC)beta1. To probe underlying mechanism(s), we developed an in vivo, immunoblot-based proteinase K (PK) protection assay to assess the changes in the P-glycoprotein content of the BBB's luminal membrane. Infusion of PK into the brain vasculature selectively cleaved luminal membrane P-glycoprotein, leaving intracellular proteins intact. Intracerebroventricular injection of VEGF partially protected P-glycoprotein from proteolytic cleavage, consistent with transporter internalization. Activation of PKCbeta1 did not protect P-glycoprotein. Thus, VEGF and PKCbeta1 reduce P-glycoprotein activity by distinct mechanisms.
Sigma-Aldrich
dPPA, ≥98% (HPLC)