GC Analysis of Lavender Flower Essential Oil on SLB®-IL60
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
oven
50 °C, 3 °C/min to 270 °C (5 min)
inj. temp.
270 °C
detector
FID, 270 °C
carrier gas
helium, 30 cm/s
injection
0.5 μL 50:1 split
sample
Lavender flower essential oil
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany