MilliporeSigma
GC Analysis of Cinnamomum camphora Essential Oil on SLB®-5ms
GC Analysis of Cinnamomum camphora Essential Oil on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C, 3 °C/min to 250 °C (5 min)
inj. temp.
280 °C
detector
FlD, 280 °C
carrier gas
helium, 30 cm/sec, constant
injection
1 μL, 50:1 split
sample
cinnamomum camphora essential oil diluted 1:10 in n-hexane
Description
Analysis Note
Cinnamomum Camphor, also known as the camphor tree, is grown in several parts of the world including Japan, Taiwan, and Australia. Camphor, which is used in medicinal and cosmetic preparations, is derived from the leaves and wood of this tree. This application illustrates the GC analysis of the volatiles of cinnamomum camphor on the SLB-5ms.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany