MilliporeSigma
GC Analysis of Fragrance Compounds in Shampoo Base on a 100% Methyl Column after SPME using 100 μm PDMS Fiber
GC Analysis of Fragrance Compounds in Shampoo Base on a 100% Methyl Column after SPME using 100 μm PDMS Fiber suitable for GC, application for SPME
Materials
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Polydimethylsiloxane (PDMS)
df 100 μm(PDMS, needle size 24 ga, for use with manual holder
CONDITIONS
SPME fiber
polydimethylsiloxane, 100 μm film (57300-U)
extraction
headspace, 5 min, 80 °C (with sample stirring)
desorption process
5 min, 250 °C
desorption temp.
-50 °C w/liquid CO2
column
100% methyl polysiloxane, 50 m × 0.25 mm I.D., 0.25 μm (Supelco equivalent, Equity-1, 28047-U)
oven
35 °C to 240 °C at 3 °C/min
inj. temp.
split, 250 °C
detector
MSD, scan m/z = 29-350 AMU
carrier gas
helium, 0.5 mL/min
sample
0.5 g shampoo base with 1% fragrance test mix, diluted 1:1 with NaCl/water in a 10 mL vial