MilliporeSigma
GC Analysis of Native Spearmint Essential Oil on SLB®-5ms
GC Analysis of Native Spearmint Essential Oil on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
75 °C (8 min), then 4 °C/min to 200 °C (15 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL, 100:1 split
liner
4 mm I.D. cup injector
sample
Native Spearmint Oil, (A.M. Todd Sample), neat
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany