MilliporeSigma
HPLC Analysis of Benoxaprofen Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Benoxaprofen Enantiomers on Astec® CHIROBIOTIC® V application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 0.1 wt% triethylamine, pH 7.0; [B] THF; (90:10, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
2 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC