MilliporeSigma
HPLC Analysis of Phenols on SUPELCOSIL LC-8
HPLC Analysis of Phenols on SUPELCOSIL™ LC-8 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-8 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
SUPELCOSIL LC-8, 15 cm × 4.6 mm I.D., 5 μm particles (58220-U)
mobile phase
[A] 1% acetic acid/methanol: [B] 1% acetic acid/water
gradient
35:65 to 100:0 (20 min), return to 35:65 (5 min)
flow rate
1.5 mL/min
column temp.
30 °C
detector
UV, 280 nm
Description
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC