MilliporeSigma
LC/MS Analysis of Amino Acids on Ascentis® Express HILIC
LC/MS Analysis of Amino Acids on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Tryptophan
Pharmaceutical Secondary Standard; Certified Reference Material
Methionine
European Pharmacopoeia (EP) Reference Standard
Tyramine
analytical standard
Tryptamine
analytical standard
Alanine
European Pharmacopoeia (EP) Reference Standard
β-Alanine
analytical standard
CONDITIONS
column
Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 μm particles (53981-U)
mobile phase
[A] 13 mM ammonium acetate in 10:90 (v/v) water: acetonitrile; [B] 13 mM ammonium acetate in water
gradient
held at 0% B for 1 min; 0 to 90% B in 19 min
flow rate
1.0 mL/min
column temp.
35 °C
detector
ESI(+)
injection
10 μL
sample
10 mg/L in Mobile Phase A
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany