MilliporeSigma
GC Analysis of Alcohols on PAG
GC Analysis of Alcohols on PAG suitable for GC
CONDITIONS
column
PAG, 30 m x 0.25 mm I.D., 0.25 μm (24223)
oven
60 °C
detector
FID, 260 °C
carrier gas
helium, 20 cm/sec
injection
1 μL
sample
0.5 - 17 mM alcohols in water