MilliporeSigma
GC Analysis of Methane, Acetylene, and Ethylene on Carboxen®-1006 PLOT and a Packed Column (Carboxen 1004)
GC Analysis of Methane, Acetylene, and Ethylene on Carboxen®-1006 PLOT and a Packed Column (Carboxen 1004) suitable for GC
CONDITIONS
column
Carboxen-1006 PLOT, 30 m x 0.53 mm I.D. (25461)
column
2m x 1/16in. stainless steel packed with Carboxen® 1004 (12846)
oven
165 °C
inj. temp.
230 °C
detector
FID, 230°C
carrier gas
helium, 22 cm/sec
injection
10 μL direct
sample
ethylene
Description
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany