HPLC Analysis of Vitamins, Water Soluble on Discovery® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] 50 mM potassium phosphate, dibasic, pH 4.5 to methanol: [B] 50mM potassium phosphate, dibasic, pH 4.0 (30:70, A:B) in 8 min
flow rate
1 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
1 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany