Skip to Content
Merck
All Photos(1)

Key Documents

6.09141

Supelco

Sodium hydroxide solution

c(NaOH) = 0.1 mol/l (0.1N solution)

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352316

Product Name

Sodium hydroxide solution, c(NaOH) = 0.1 mol/l (0.1N solution)

vapor pressure

3 mmHg ( 37 °C)

form

solution

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Development of biodegradable film from cactus (Opuntia Ficus Indica) mucilage loaded with acid-leached kaolin as filler.: This study highlights the application of sodium hydroxide solution in the development of biodegradable films using cactus mucilage. The process enhances the film′s mechanical properties and demonstrates an innovative use of natural resources in sustainable material science (Teshager AA et al., 2024).

  • Basic research on layered geopolymer composites with insulating materials of natural origin.: Sodium hydroxide solution plays a crucial role in the synthesis of geopolymer composites, contributing to the enhancement of thermal insulation properties. This research provides insights into the use of geopolymers in building materials, focusing on sustainability and energy efficiency (Bąk A et al., 2024).

  • Potential for Recycling Metakaolin/Slag-Based Geopolymer Concrete of Various Strength Levels in Freeze-Thaw Conditions.: The use of sodium hydroxide solution in the preparation of metakaolin/slag-based geopolymers shows significant resistance to freeze-thaw cycles, suggesting its utility in creating durable building materials suitable for cold climates (Liu M et al., 2024).

  • Preparation of nickel foam modified by multiwalled hollow spheres of NiCo(2)O(4) as a promising non-enzymatic glucose sensor.: This research utilizes sodium hydroxide solution to modify nickel foam, enhancing its application in non-enzymatic glucose sensing. The study advances the development of cost-effective and highly sensitive biosensors (Eprilia N et al., 2024).

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service