Skip to Content
Merck
All Photos(1)

Key Documents

6.09956

Supelco

Sodium hydroxide solution

c(NaOH) = 1 mol/l (1N), concentrate for preparing 0.5 L volumetric solution for titration, Titriplex®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352316

Product Name

Sodium hydroxide solution, for 500 ml, c(NaOH) = 1 mol/l (1N) Titrisol

grade

reagent

vapor pressure

3 mmHg ( 37 °C)

form

solution

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Characterization and biological activities of polysaccharides extracted from Auricularia auricula with different extraction methods.: This study demonstrates the use of sodium hydroxide solution in optimizing the extraction of biologically active polysaccharides from Auricularia auricula. The research provides insight into how varying extraction methods affect the yield and functionality of polysaccharides, which is critical for both food science and pharmaceutical applications (He G et al., 2023).


  • Extraction process, physicochemical properties, and digestive performance of red yeast rice starch.: The application of sodium hydroxide solution in the extraction process of starch from red yeast rice highlights its role in modifying starch properties and enhancing its digestibility. This research is vital for developments in food processing and nutritional science (Bai M et al., 2024).


  • [Determination of boric acid and silicic acid in mineral water by nonsuppressed ion chromatography].: This study utilizes sodium hydroxide solution for the chromatographic analysis of mineral waters, underscoring its importance in environmental analytical chemistry for ensuring water quality and safety (Yang Z et al., 2023).


  • Chitosan-modified cotton fiber: An efficient and reusable adsorbent in removal of harmful cyanobacteria, Microcystis aeruginosa from aqueous phases.: This article explores the role of sodium hydroxide solution in modifying cotton fibers with chitosan for the removal of cyanobacteria from water. It highlights the potential for developing effective and sustainable water treatment technologies (Park YH et al., 2024).


Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service