Skip to Content
Merck
All Photos(1)

Key Documents

6.17692

Supelco

Buffer capsules

pH 4.00±0.05

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

pH

4.00±0.05

Application


  • Comparison of Two Grafted Copolymers, Soluplus and Kollicoat IR, as Solid Dispersion Carriers of Arteether for Oral Delivery Prepared by Different Solvent-Based Methods.: This study compares the efficacy of two grafted copolymers in buffer capsules for oral delivery of Arteether, demonstrating significant improvements in drug solubility and bioavailability (Desai and Chatterjee, 2023).

  • Retrospective analysis of the biopharmaceutics characteristics of solid oral Modified-Release drug products in approved US FDA NDAs designated as Extended-Release or Delayed-Release formulations.: This analysis highlights the biopharmaceutical properties of buffer capsules in modified-release drug formulations, providing insights into their effectiveness and regulatory approval processes (Ahmed et al., 2023).

  • Crushing the Contents of Valbenazine Capsules for Potential Addition to Soft Foods or Administration via Gastrostomy Tube.: The study evaluates the feasibility of crushing valbenazine buffer capsules for alternative administration routes, ensuring drug stability and patient compliance (Sajatovic et al., 2023).


Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service