6.17693
Buffer capsules
pH 7.00±0.05
Synonym(s):
Buffer Pill
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161706
Recommended Products
pH
7.00±0.05
Application
- Engineering 5-flourouracil and leucovorin-loaded vesicular systems for possible colon specific delivery: In vitro evaluation and real time cell assay against HCT-116 colon cell lines.: This study explores the use of vesicular systems to deliver chemotherapy agents specifically to the colon, utilizing buffer capsules to stabilize the drug formulation. The findings suggest enhanced efficacy and targeted delivery, promising advancements in colon cancer treatment (Ugorji et al., 2024).
- Effect of hydroxypropyl methylcellulose (HPMC) hard capsules on in vitro rumen fermentation kinetics of commonly used feeds in ruminant feeding.: This study investigates the impact of HPMC hard buffer capsules on the fermentation kinetics in ruminant feeds, providing insights into improving feed efficiency and digestion in livestock management (Valcl and Lavrenčič, 2024).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service