Skip to Content
Merck
All Photos(1)

Key Documents

6.18608

Supelco

Silica gel

for column chromatography (100-200 mesh)

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352309

grade

for column chromatography (100-200 mesh)

bp

2230 °C

mp

>1600 °C

suitability

suitable for column chromatography

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Preparation and chromatographic evaluation of 9-oxa-10-phosphophenanthrene 10-oxide bonded phenyl stationary phase and investigation of its retention mechanism through nuclear magnetic resonance spectroscopy.: This study discusses the innovative preparation of a silica gel-based chromatographic stationary phase, specifically designed to enhance the selectivity and stability of the chromatographic process. The unique bonding of 9-oxa-10-phosphophenanthrene 10-oxide to a phenyl stationary phase offers insights into the molecular interactions involved, enhancing the analytical chemist′s ability to segregate and identify chemical species efficiently (Xiao et al., 2024).
  • Green synthesis of N-rich carbon dot-derived crosslinked covalent organic nanomaterial for multipurpose chromatographic applications.: This research introduces a green synthetic route to develop nitrogen-rich carbon dots, which are then utilized to create crosslinked covalent organic frameworks. These frameworks are applied as novel stationary phases in chromatography, showcasing superior performance in terms of thermal stability and chemical inertness, making them ideal for complex analytical tasks in academia and beyond (Wei et al., 2024).
  • Isomerization of E-Cinnamamides into Z-Cinnamamides Using a Recycling Photoreactor.: This paper presents a novel method using silica gel as a support medium in a recycling photoreactor for the isomerization of E-cinnamamides into Z-cinnamamides. The process highlights the material′s role in facilitating photochemical reactions, providing a sustainable and efficient approach to molecular transformations critical in synthetic chemistry (Suga et al., 2024).
  • Antibacterial and Anti-Inflammatory Properties of Flavonoids from Streptomyces chartreusis RH3.5.: Although primarily focused on biological extracts, this study utilizes silica gel for the purification and analysis of antibacterial and anti-inflammatory compounds. This emphasizes silica gel′s crucial role in the extraction and refinement processes in pharmaceutical research, underpinning its utility in developing therapeutic agents (Taechowisan et al., 2024).
  • Cytotoxic Sulfoquinovosyl Glycerols from the Seaweed Sargassum Angustifolium from Persian Gulf.: This investigation employs silica gel chromatography to isolate and identify cytotoxic compounds from seaweed, demonstrating the adaptability of silica gel in handling diverse samples and contributing to marine biomedicine research. This application underscores its indispensable role in the extraction and purification of bioactive molecules (Sajjadi et al., 2024).

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service