Skip to Content
Merck
All Photos(1)

Key Documents

6.18609

Supelco

Silica gel

for column chromatography (230-400 mesh)

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352309

grade

for column chromatography (230-400 mesh)

bp

2230 °C

mp

>1600 °C

suitability

suitable for column chromatography

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • A sulfated exopolysaccharide derived from Chlorella sp. exhibiting in vitro anti-α-D-Glucosidase activity: Sulfuric acid′s role in the synthesis and modification of biochemical compounds is demonstrated, underscoring its utility in pharmaceutical research and development (Guehaz et al., 2024).

  • Emission characteristics of condensable particulate matter during the production of solid waste-based sulfoaluminate cement: This study uses sulfuric acid to analyze environmental pollutants, reflecting its importance in environmental science for testing and analysis (Li et al., 2024).




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service