Skip to Content
Merck
All Photos(1)

Key Documents

71755-M

Sigma-Aldrich

2xYT Broth - Novagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
NACRES:
NA.85

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-30°C

General description

2xYT Broth is a powdered medium for the enrichment of E. coli. The composition of the 2xYT Broth per liter is 16 g tryptone, 10 g yeast extract, and 5 g sodium chloride.

Warning

Toxicity: Standard Handling

Other Notes

Not available for sale in Austria, Belgium, Denmark, Finland, France, Germany, Ireland, Italy, Luxembourg, Netherlands, Norway, Portugal, Spain, Sweden, Switzerland, and UK.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service