Skip to Content
Merck
All Photos(1)

Key Documents

SRE0118

Sigma-Aldrich

dsDNase

recombinant, expressed in Pichia pastoris, heat-labile, Triton-free

Synonym(s):

double strand specific deoxyribonuclease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in Pichia pastoris

Quality Level

specific activity

(Ca. 400 000 Kunitz Units/mg)

feature

heat-labile

composition

Triton-free

packaging

pkg of 250 U

optimum pH

7.5

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

dsDNase is a heat-labile endonuclease that has a strong preference for double-stranded DNA. dsDNase can be heat inactivated by heat treatment at 15 min at 65˚C, or 20 min at 60˚C. The enzyme requires 1 mM DTT and pH ≥ 8 for complete inactivation.

Application

This dsDNase is a useful tool to remove contaminating DNA from PCR mastermixes.

Features and Benefits

  • This nuclease has double-stand specificity, which allows specific degradation of dsDNA while leaving shorter ssDNA as primers.
  • Easy inactivation by moderate heat (65°C) allows addition of DNA intended for analysis directly after removal of contaminating DNA.
  • The specific activity is estimated to be 30 times higher than that of bovine DNase I.
Activity towards dsDNA is 5000-fold higher than towards ssDNA

Unit Definition

One Unit is defined as an increase in absorbance at 260 nm of 0.001 per minute, using 50 µg/ml high MW DNA in 100 mM Na-acetate pH 5.0 and 5 mM MgCl2.

Reconstitution

In solution.

Other Notes

The dsDNase is highly active in a temperature range of 20-40°C. It needs at least 2.5 mM Mg for activity and has an optimal pH at 7.5.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service