SRE0119
dsDNase
For gDNA removal, recombinant, expressed in Pichia pastoris, Heat labile, Triton-free
Synonym(s):
HL-dsDNase, heat labile double strand specific deoxyribonuclease
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.77
Recommended Products
recombinant
expressed in Pichia pastoris
Quality Level
composition
Triton-free
packaging
pkg of 250 U
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Double-strand specific DNases (dsDNases) are unique double-strand specific endonucleases. As they do not digest ssDNA or RNA, they can be used to specifically remove dsDNA in the presence of other nucleic acids.
Application
This heat labile dsDNase removes genomic DNA from RNA preparations with minimal impact on RNA integrity.with minimal impact on RNA integrity
Features and Benefits
Due to the mild inactivation temperature required, this dsDNase can remove genomic DNA from RNA perparations and have minimal impact onto RNA quality and quantity.
Unit Definition
One unit is defined as an increase in absorbance at 260 nm of 0.001 per minute, using 50 µg/ml of high MW DNA in 100 mM Na-acetate pH 5.0 and 5 mM MgCl2.
Reconstitution
In solution
Other Notes
The heat labile dsDNase is highly active in a temperature range of 20-40°C. It needs at least 2.5 mM Mg for activity and has an optimal pH at 7.5.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service