Skip to Content
Merck
All Photos(1)

Key Documents

880242P

Avanti

DMG-PEG(2K)-DBCO

Synonym(s):

1,2-dimyristoyl-rac-glycero-3-[N-dibenzocyclooctyl(polyethylene glycol)-2000]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C140H254N2O51
CAS Number:
Molecular Weight:
2781.50
UNSPSC Code:
12352200

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (880242P-25MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

-10 to -25°C

General description

One of the challenges associated with LNP′s is effectively delivering the payload to the target cells. Click chemistry presents a promising solution for the straightforward attachment of functional groups to other molecules, enabling the synthesis of larger and more complex molecular structures. Clickable PEG lipids allow for the easy linkage of specific cell-targeting functional groups to the PEG lipid, enhancing delivery to the intended cells. Additionally, by adjusting the PEG length, one can modify micelle size, immunological safety, and drug release rates.

Application

Targeted Drug Delvery

Packaging

5 mL Amber Glass Screw Cap Vial (880242P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service