Skip to Content
Merck
All Photos(1)

Key Documents

880251S

Avanti

PEI HCl

Avanti Research - A Croda Brand

Synonym(s):

Linear Polyethyleneimine (LPEI) for High-Efficiency Viral Vector Production, Polyethylenimine (PEI) hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9NO
Molecular Weight:
75.11
MDL number:
UNSPSC Code:
12352200

biological source

synthetic

Assay

≥99% (TLC)

form

aqueous solution

packaging

pkg of 1 × 1 L (880251S-1G)
pkg of 1 × 1 mL (880251S-1.5MG)
pkg of 1 × 10 mL (880251S-10MG)
pkg of 1 × 100 mL (880251S-100MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CNCCO

InChI

1S/C3H9NO/c1-4-2-3-5/h4-5H,2-3H2,1H3

InChI key

OPKOKAMJFNKNAS-UHFFFAOYSA-N

General description

Avanti Research′s Linear Polyethyleneimine (LPEI) is a premium transfection reagent specifically engineered for effective viral vector production in both adherent and suspension cell culture systems. Renowned for its capability to create stable complexes with DNA or RNA, LPEI improves cellular uptake and guarantees high transfection efficiency. This makes it an excellent option for researchers engaged in gene therapy, vaccine development, and biomanufacturing.

Application

Vaccine development

Packaging

1.5 mL polypropylene microtube (880251S-1.5MG), 20 mL PETG Square Bottle (880251S-10MG), 125 mL PETG Square Bottle (880251S-100MG), 1000 mL PETG Square Bottle (880251S-1G)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service