Skip to Content
Merck
All Photos(1)

Key Documents

SBR00060

Sigma-Aldrich

Bedaquiline Fumarate

≥97% (HPLC), powder

Synonym(s):

Baloxin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C32H31BrN2O2·C4H4O4
CAS Number:
UNSPSC Code:
51101500

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to off-white

antibiotic activity spectrum

mycobacteria

Mode of action

enzyme | inhibits

SMILES string

CN(C)CC[C@@](C1=CC=CC2=CC=CC=C21)([C@H](C3=CC=CC=C3)C4=C(N=C5C=CC(=CC5=C4)Br)OC)O.C(=C/C(=O)O)\C(=O)O

InChI

1S/C32H31BrN2O2.C4H4O4/c1-35(2)19-18-32(36,28-15-9-13-22-10-7-8-14-26(22)28)30(23-11-5-4-6-12-23)27-21-24-20-25(33)16-17-29(24)34-31(27)37-3;5-3(6)1-2-4(7)8/h4-17,20-21,30,36H,18-19H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t30-,32-;/m1./s1

InChI key

ZLVSPMRFRHMMOY-WWCCMVHESA-N

General description

Bedaquiline fumarate is an antibioitc with antimycobacterial properties. Bedaquiline fumarate a fumarate salt prepared from equimolar amounts of bedaquiline and fumaric acid. It has a role as an antitubercular agent and an ATP synthase inhibitor.

Application

Bedaquiline Fumarate has also been studied for its potential use in bioluminescence imaging, due to its ability to bind to a reporter protein and enhance its luminescence signal. This has potential applications in a variety of biological research areas, such as protein-protein interactions, drug discovery, and disease modeling.

Biochem/physiol Actions

Bedaquiline Fumarate is an antimycobacterial agent that inhibits mycobacterial ATP synthase, an enzyme essential for energy metabolism in Mycobacterium tuberculosis. Inhibition of ATP synthase disrupts the energy production of the bacteria and leads to their death. Bedaquiline Fumarate is active against mycobacteria.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service