주요 문서
Z740472
MICROGARD 2000 coveralls for protection from infective agent
size XL (elasticated ankles)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
46181503
NACRES:
NB.35
크기
XL (elasticated ankles)
제조업체/상표
Arco 1762006
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
MICROGARD 2000 fully elasticated coverall with hood , setting the standard in Type 5/6 performance and protection, offers protection from infective agents, pesticides, particulates, fibres and liquid chemical spray.
Features:
  • 3-piece hood
  • 2-way front zipper with re-sealable storm flap
  • Finger loops
  • Elasticated hood, waist, wrists and ankles
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-5 Anti-static
법적 정보
Microgard is a trademark of Microgard Limited
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)