주요 문서
Z740483
MICROGARD 2500 coveralls for protection from saturating liquid spray
size XL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
46181503
NACRES:
NB.35
크기
XL
제조업체/상표
Arco 1724206
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
MICROGARD 2500 STANDARD is designed to protect from saturating liquid spray and particulates, the fabric′s physical strength and flexibility ensure you are protected yet comfortable.
Features:
  • 3 piece hood
  • Ultrasonically welded seams
  • Finger loops
  • 2-way front zip with resealable storm flap
  • Elasticated hood, waist, wrists and ankles
Approved to:
EN14605 Type 4 Spray-tight
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-1 Anti-static
법적 정보
Microgard is a trademark of Microgard Limited
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)