MABN640-I
Anti-Amyloid beta fibrils Antibody, clone M98
clone M98
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
Recommended Products
clone
M98, recombinant monoclonal
isotype
IgG
Specificity
Clone M98 is a recombinant rabbit monoclonal antibody that detects Amyloid beta fibrils. It recognizes a specific conformation of residues 2-7 of the amyloid beta peptide sequence (AEFRHD) found in amyloid fibrils.
Immunogen
Fibrillar Abeta 1-42 peptide
Reconstitution
Please refer to lot specific datasheet.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service