Skip to Content
Merck
All Photos(1)

Key Documents

PHL86281

β-Crocetin

phyproof® Reference Substance

Synonym(s):

8,8′-Diapo-ψ,ψ-carotenedioic acid monomethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O4
CAS Number:
Molecular Weight:
342.43
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

storage temp.

−20°C

SMILES string

O=C(/C(C)=C/C=C/C(C)=C/C=C/C=C(/C=C/C=C(C(O)=O)\C)C)OC

InChI

1S/C21H26O4/c1-16(12-8-14-18(3)20(22)23)10-6-7-11-17(2)13-9-15-19(4)21(24)25-5/h6-15H,1-5H3,(H,22,23)/b7-6+,12-8+,13-9+,16-10+,17-11+,18-14+,19-15+

InChI key

DGGQUKOLHQXDLV-QTNXRKSWSA-N

Related Categories

General description

This Substance belongs to the class of carotenoids.
This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service