Skip to Content
Merck
All Photos(1)

Key Documents

V800067

Sigma-Aldrich

Barium nitrate

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(NO3)2
CAS Number:
Molecular Weight:
261.34
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
AR
form:
solid

grade

AR

product line

Vetec

Assay

≥99.5%

form

solid

pH

5-8 (5% in water)

mp

592 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.0005%

cation traces

Fe: ≤0.0002%
K: ≤0.005%
Sr: ≤0.05%

SMILES string

[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ba.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

IWOUKMZUPDVPGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service