Key Documents
AATB-21166
AAT Bioquest
AAT Bioquest Coelenterazine
10 mg
About This Item
CAS Number:
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
AAT Bioquest
General description
Native coelenterazine and its derivatives confer different Ca2+ affinities and spectral properties on the aequorin complex. Recombinant apoaequorin reconstituted with coelenterazine hcp is reported to have the best luminescence overall, with both a high quantum yield and a fast response time. However, intracellular reconstitution of aequorin from coelenterazine analogs can be relatively slow. Aequorins containing the cp, f or h form of coelenterazine exhibit 10-20 times stronger luminescence than that of apoaequorin reconstituted with native coelenterazine. Coelenterazines h and cp has been used in HTS screening assay for GPCRs.
Coelenterazine *CAS#: 55779-48-1* is manufactured by AAT Bioquest
Choose from one of the most recent versions:
Certificates of Analysis (COA)