50973
TrippNT
TrippNT® HPLC 30 Column PVC Cabinet
White PVC and Clear Acrylic with 30 Racks and Double Hinged Acrylic Doors
Synonym(s):
cabinet, storage, hplc organization
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
material
PVC
weight
3.58 lb
manufacturer/tradename
TrippNT
Related Categories
General description
TrippNT's HPLC 60 Column PVC Cabinet with Lockable Hinged Doors protects, stores and organizes columns. This storage cabinet allows for inventory to be easily seen and features a hasp lock. Is wall mounted using keyholes and included mounting hardware. All TrippNT products are made in the USA and come with a 100% Total Replacement Lifetime Guarantee.
- Rectangular Design with 30 Racks and Double Hinged Acrylic Doors
- Hasp Lock for Doors
- Constructed from White PVC and Clear Acrylic
- Designates Safe Place for Columns
- Finish: Smooth, no rough or sharp edges
- Material: 1/4" White PVC, 1/8" Clear Acrylic
- Product Weights: 3.58 lbs
- Ships from: Kansas City, MO, USA
Legal Information
TrippNT® is a registered trademark of TrippNT, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.