MilliporeSigma
HPLC Analysis of Acebutolol Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Acebutolol Enantiomers on Astec® CHIROBIOTIC® T2 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T2 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine; (99.8:0.1:0.1, A:B:C)
flow rate
1 mL/min
column temp.
22 °C
detector
UV, 254 nm
injection
12 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC